missing translation for 'onlineSavingsMsg'
Learn More

Centrophenoxine hydrochloride, 98%

Catalog No. AAJ6419606
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6419606 5 g
AAJ6419614 25 g
AAJ6419622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6419606 Supplier Thermo Scientific Chemicals Supplier No. J6419606

CAS: 3685-84-5 | C12H17Cl2NO3 | 294.17 g/mol

Ester of dimethylethanolamine and 4-chlorophenoxyacetic acid; activates acetylcholinesterase activity in hippocampus of aged rats

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ester of dimethylethanolamine and 4-chlorophenoxyacetic acid; activates acetylcholinesterase activity in hippocampus of aged rats

Chemical Identifiers

CAS 3685-84-5
Molecular Formula C12H17Cl2NO3
Molecular Weight (g/mol) 294.17
MDL Number MFCD00012533
InChI Key FIVHOHCAXWQPGC-UHFFFAOYSA-N
Synonym meclofenoxate hydrochloride, centrophenoxine hydrochloride, lucidril, cerutil, meclofenoxate hcl, atsefen, helfergin, marucotol, acefen, brenal
PubChem CID 19379
IUPAC Name hydrogen 2-(dimethylamino)ethyl 2-(4-chlorophenoxy)acetate chloride
SMILES [H+].[Cl-].CN(C)CCOC(=O)COC1=CC=C(Cl)C=C1

Specifications

Melting Point 135°C to 145°C
Color White
Quantity 5 g
Merck Index 14,5780
Formula Weight 294.17
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Centrophenoxine hydrochloride
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 222-975-3
RTECSNumber AG0440000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only