missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Cellulose Nitrate Membrane Filter
GREENER_CHOICE

Catalog No. 14001948
Change view
Click to view available options
Pore Size:
0.45 μm
0.8 μm
1.2 μm
5 μm
8 μm
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pore Size
14001948 5 μm
14001953 0.45 μm
14001956 0.8 μm
14001959 1.2 μm
14001963 8 μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 14001948 Supplier Sartorius Supplier No. 11342293G
Only null left

Cellulose nitrate membrane filters with 293 mm diameters are hydrophilic, have high flow rates thanks to their symmetrical structure and are compatible with aqueous solutions, hydrocarbons and several other organic solvents.

  • Ideal for several general laboratory applications
  • High, non-specific absorption
  • Hydrophilic, with high flow rates
  • 25 filters
  • 293 mm diameter

Specifications

Disposable Single-use
Filter Type Membrane Filter
Format Disc
Material Cellulose Nitrate
Pore Size 5 μm
Sterility Non-sterile
Quantity 25/Cs.
Type Membrane Filter
Diameter (Metric) 293 mm