Learn More
Invitrogen™ Cells-to-CT™ Bulk Fast Advanced RT Reagents
Invitrogen Fast Advanced Cells-to-CT Bulk RT Reagents are separately-available, large-quantity versions of the reagents used for reverse transcription (RT) in the TaqMan Fast Advanced Cells-to-CT Kit and the SYBR Green Fast Advanced Cells-to-CT Kit.
Supplier: Invitrogen™ A39110
Comprend: Cells-to-Ct 20X Fast Advanced RT enzyme mix and 2X RT buffer
Description
Invitrogen Fast Advanced Cells-to-CT Bulk RT Reagents are separately-available, large-quantity versions of the reagents used for reverse transcription (RT) in the TaqMan Fast Advanced Cells-to-CT Kit and the SYBR Green Fast Advanced Cells-to-CT Kit (20X RT enzyme mix and 20X RT buffer).
Spécifications
| 20X | |
| Primer-probe | |
| Fast | |
| High-throughput Compatible | |
| Bulk Fast Advanced Rt Reagent | |
| Dry Ice | |
| Gene Expression | |
| 2500 Reactions | |
| Cell |
| • 6.8 mL 20X Fast Advanced RT Enzyme Mix • 68 mL 2X Fast Advanced RT Buffer Store at -5° to -30°C. |
|
| High | |
| Fewer resources used and less waste, Less hazardous | |
| Cells-to-CT | |
| 2500 reactions | |
| 2500 Reactions | |
| 20X | |
| Lysate | |
| 68 mL |
For Research Use Only. Not for use in diagnostic procedures.