Learn More
Corning™ Cell Recovery Solution
GREENER_CHOICE
Allows for the recovery of cells cultured on Corning™ Matrigel™ Basement Membrane Matrix for subsequent biochemical analyses
$216.89 - $741.65
Specifications
Content And Storage 2°C to 8°C
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
08774405
GREENER_CHOICE
View Documents Promotion Details
Corning™
354270
500 mL
Each for $741.65
Only null left
CB40253
GREENER_CHOICE
View Documents Promotion Details
Corning™
354253
100 mL
Each for $216.89
Only null left
Description
Description
Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.
Specifications
Specifications
2°C to 8°C