missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Cell Proliferation Dye eFluor™ 450

A violet fluorescent dye that can be used to monitor individual cell divisions for up to 7 generations.

$201.00 - $590.00

Specifications

Color Violet
Detection Method Fluorescence
For Use With (Application) Proliferation Assay
For Use With (Equipment) Flow Cytometer
Product Type Dye
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50246096
View Documents Promotion Details
Invitrogen™
65084285
500 μg
Each for $201.00
Only null left
 
501128735
View Documents Promotion Details
Invitrogen™
65084290
4 x 500 μg
Each for $590.00
Only null left
 
Description

Description

  • A violet fluorescent dye that can be used to monitor individual cell divisions
  • Dye binds to any cellular proteins containing primary amines, and as cells divide, the dye is distributed equally between daughter cells
  • They can be measured as successive halving of the fluorescence intensity of the dye - visualize up to 7 generations
  • Cells labeled with Cell Proliferation Dye eFluor™ 450 may be fixed and permeabilized for analysis of intracellular targets using standard formaldehyde-containing fixatives and saponin-based permeabilization buffers
  • Cell Proliferation Dye eFluor™ 450 has a peak excitation of 409 nm and can be detected using a 450/50 band pass filter (equivalent to eFluor™ 450 or Pacific Blue™), making it compatible with applications that utilize GFP

Additional Details

  • Each vial may be reconstituted to a stock concentration of 10 mM with 165 μL of anhydrous DMSO
  • Once reconstituted it should be used within 6 months and protected from light and stored at -20°C with dessicant
  • Avoid freeze-thawing
Specifications

Specifications

Violet
Proliferation Assay
Dye
Lyophilized powder
Fluorescence
Flow Cytometer
405/450
eBioscience
Videos
SDS
Documents

Documents

For Research Use Only. Not for use in diagnostic procedures.