missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Celecoxib

Catalog No. 378610
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
378610 10 mg
378650 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 378610 Supplier Tocris Bioscience™ Supplier No. 3786/10

Selective cyclooxygenase-2 (COX-2) inhibitor

Celecoxib is a selective cyclooxygenase-2 (COX-2) inhibitor (IC50 values are 15 and 0.04 μM for COX-1 and COX-2 respectively). Celecoxib is an anti-inflammatory with shorter plasma half-life in vivo than SC 58121. Displays chemopreventive activity in in vivo tumor models.

Chemical Identifiers

CAS 169590-42-5
Molecular Formula C17H14F3N3O2S

Specifications

Quantity 10 mg
Formula Weight 381.37
Purity >99%
Molecular Weight (g/mol) 381.373
Chemical Name or Material Celecoxib
Recommended Storage Store at Room Temperature