missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Celastrol
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Quantity | 100 mg |
| Melting Point | 203.0°C |
| Formula Weight | 450.62 |
| InChI Key | KQJSQWZMSAGSHN-JJWQIEBTSA-N |
| SMILES | CC1=C(O)C(=O)C=C2C1=CC=C1[C@@]2(C)CC[C@@]2(C)[C@@H]3C[C@@](C)(CC[C@]3(C)CC[C@]12C)C(O)=O |
| IUPAC Name | (2R,4aS,6aS,12bR,14aS,14bR)-10-hydroxy-2,4a,6a,9,12b,14a-hexamethyl-11-oxo-1,2,3,4,4a,5,6,6a,11,12b,13,14,14a,14b-tetradecahydropicene-2-carboxylic acid |
| Molecular Weight (g/mol) | 450.62 |
| Chemical Name or Material | Celastrol |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Toxic if swallowed. |
| Precautionary Statement | GHS P Statement Wash face, hands and any exposed skin thoroughly after handling. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Store locked up. |
RUO – Research Use Only