Learn More
Cefmetazole sodium, 921μg/mg, Thermo Scientific Chemicals
$177.58 - $177.58
Chemical Identifiers
| CAS | 56796-39-5 |
|---|---|
| Molecular Formula | C15H16N7NaO5S3 |
| Molecular Weight (g/mol) | 493.507 |
| MDL Number | MFCD00214260 |
| InChI Key | BITQGIOJQWZUPL-PBCQUBLHSA-M |
| PubChem CID | 23666711 |
| ChEBI | CHEBI:3490 |
| IUPAC Name | sodium;(6R,7S)-7-[[2-(cyanomethylsulfanyl)acetyl]amino]-7-methoxy-3-[(1-methyltetrazol-5-yl)sulfanylmethyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate |
| SMILES | CN1C(=NN=N1)SCC2=C(N3C(C(C3=O)(NC(=O)CSCC#N)OC)SC2)C(=O)[O-].[Na+] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAJ6634803
|
Thermo Scientific Chemicals
J6634803 |
1 g |
Each for $177.58
|
|
|||||
Description
Cefmetazole is used to study the effect of expression, binding, and inhibition of penicillin-binding proteins (with the exception of PBP2) on bacterial cell wall mucopeptide synthesis. It is also used to study protein mediated antibiotic transport.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Cefmetazole is used to study the effect of expression, binding, and inhibition of penicillin-binding proteins (with the exception of PBP2) on bacterial cell wall mucopeptide synthesis
- It is also used to study protein mediated antibiotic tr
Chemical Identifiers
| 56796-39-5 | |
| 493.507 | |
| BITQGIOJQWZUPL-PBCQUBLHSA-M | |
| CHEBI:3490 | |
| CN1C(=NN=N1)SCC2=C(N3C(C(C3=O)(NC(=O)CSCC#N)OC)SC2)C(=O)[O-].[Na+] |
| C15H16N7NaO5S3 | |
| MFCD00214260 | |
| 23666711 | |
| sodium;(6R,7S)-7-[[2-(cyanomethylsulfanyl)acetyl]amino]-7-methoxy-3-[(1-methyltetrazol-5-yl)sulfanylmethyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate |
Specifications
| Cefmetazole Sodium | |
| 56796-39-5 | |
| 1 g | |
| C15H16N7NaO5S3 | |
| Soluble in water | |
| CN1C(=NN=N1)SCC2=C(N3C(C(C3=O)(NC(=O)CSCC#N)OC)SC2)C(=O)[O-].[Na+] | |
| 493.507 | |
| CHEBI:3490 | |
| Cefmetazole sodium, 921μg/mg |
| Powder | |
| White | |
| Hygroscopic | |
| MFCD00214260 | |
| BITQGIOJQWZUPL-PBCQUBLHSA-M | |
| sodium;(6R,7S)-7-[[2-(cyanomethylsulfanyl)acetyl]amino]-7-methoxy-3-[(1-methyltetrazol-5-yl)sulfanylmethyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate | |
| 23666711 | |
| 493.52 |
Safety and Handling
GHS H Statement
H334-H335-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
P261-P264b-P271-P272-P280-P285-P302+P352-P304+P340-P305+P351+P338-P333+P313-P342+P311-P362-P501c
H315-H317-H319-H334-H335
RTECSNumber : XI0372200
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only