accessibility menu, dialog, popup

UserName

CAS RN 56796-39-5

IUPAC Name: sodium (6R,7S)-7-{2-[(cyanomethyl)sulfanyl]acetamido}-7-methoxy-3-{[(1-methyl-1H-1,2,3,4-tetrazol-5-yl)sulfanyl]methyl}-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
Synonyms: zefazone CS 1170 sodium CMZ cmz sodium skf 83088 sodium cefmetazone cefmetazon
Molecular Weight (g/mol): 493.51
Molecular Formula: C15H16N7NaO5S3
InChi Key: BITQGIOJQWZUPL-PBCQUBLHSA-M
SMILES: [Na+].CO[C@]1(NC(=O)CSCC#N)[C@H]2SCC(CSC3=NN=NN3C)=C(N2C1=O)C([O-])=O
Physical Form 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Percent Purity 
  • (2)
Color 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results