missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cefixime, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 459480050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Cefixime | |
| 2.6 to 4.1 | |
| 97.5% min. (HPLC) | |
| MFCD00865020 | |
| cefixime, cefixima, cefiximum, cefixim, cefspan, cephoral, cefixoral, cefiximum latin, suprax, necopen | |
| NC1=NC(=CS1)C(=N\OCC(O)=O)\C(=O)N[C@H]1[C@H]2SCC(C=C)=C(N2C1=O)C(O)=O | |
| 453.44 | |
| CHEBI:472657 | |
| 98% |
| 79350-37-1 | |
| Authentic | |
| C16H15N5O7S2 | |
| 5 g | |
| OKBVVJOGVLARMR-QSWIMTSFSA-N | |
| (6R,7R)-7-[[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-(carboxymethoxyimino)acetyl]amino]-3-ethenyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid | |
| 5362065 | |
| 453.45 |
Chemical Identifiers
| 79350-37-1 | |
| 453.44 | |
| OKBVVJOGVLARMR-QSWIMTSFSA-N | |
| 5362065 | |
| (6R,7R)-7-[[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-(carboxymethoxyimino)acetyl]amino]-3-ethenyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid |
| C16H15N5O7S2 | |
| MFCD00865020 | |
| cefixime, cefixima, cefiximum, cefixim, cefspan, cephoral, cefixoral, cefiximum latin, suprax, necopen | |
| CHEBI:472657 | |
| NC1=NC(=CS1)C(=N\OCC(O)=O)\C(=O)N[C@H]1[C@H]2SCC(C=C)=C(N2C1=O)C(O)=O |
RUO – Research Use Only