accessibility menu, dialog, popup

UserName

CAS RN 79350-37-1

IUPAC Name: (6R,7R)-7-[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-[(carboxymethoxy)imino]acetamido]-3-ethenyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
Synonyms: cefixime (-)-cefixim cefiximum cefixima
Molecular Weight (g/mol): 453.44
Molecular Formula: C16H15N5O7S2
InChi Key: OKBVVJOGVLARMR-QSWIMTSFSA-N
SMILES: NC1=NC(=CS1)C(=N\OCC(O)=O)\C(=O)N[C@H]1[C@H]2SCC(C=C)=C(N2C1=O)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results