Learn More
Cefazolin sodium salt
Inhibits bacterial cell wall synthesis | CAS: 27164-46-1 | C14H16N8NaO4S3 | 479.504 g/mol
Supplier: Thermo Scientific Chemicals J6527406
Description
Inhibits bacterial cell wall synthesis
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Cefazolin Sodium Salt | |
| Powder | |
| 170°C to 172°C | |
| 5 g | |
| C14H16N8NaO4S3 | |
| 3585038 | |
| Cefamedin | |
| [HH].CC1=NN=C(S1)SCC2=C(N3C(C(C3=O)NC(=O)CN4C=NN=N4)SC2)C(=O)O.[Na] | |
| 479.504 | |
| 476.48 |
| Cefazolin sodium salt | |
| 27164-46-1 | |
| White | |
| Light sensitive | |
| MFCD00056883 | |
| 14,1917 | |
| MTIAAUXSENDLGW-SLNAEPSVSA-N | |
| (6R,7R)-3-[(5-methyl-1,3,4-thiadiazol-2-yl)sulfanylmethyl]-8-oxo-7-[[2-(tetrazol-1-yl)acetyl]amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;molecular hydrogen;sodium | |
| 131673922 |
Chemical Identifiers
| 27164-46-1 | |
| 479.504 | |
| MTIAAUXSENDLGW-SLNAEPSVSA-N | |
| 131673922 | |
| [HH].CC1=NN=C(S1)SCC2=C(N3C(C(C3=O)NC(=O)CN4C=NN=N4)SC2)C(=O)O.[Na] |
| C14H16N8NaO4S3 | |
| MFCD00056883 | |
| Cefamedin | |
| (6R,7R)-3-[(5-methyl-1,3,4-thiadiazol-2-yl)sulfanylmethyl]-8-oxo-7-[[2-(tetrazol-1-yl)acetyl]amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;molecular hydrogen;sodium |
Safety and Handling
GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
P261-P272-P280g-P285-P302+P352-P304+P341-P333+P313-P342+P311-P363-P501c
H317-H334
EINECSNumber : 248-278-4
RTECSNumber : XI0390000
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only