accessibility menu, dialog, popup

UserName

CAS RN 27164-46-1

IUPAC Name: sodium 3-{[(5-methyl-1,3,4-thiadiazol-2-yl)sulfanyl]methyl}-8-oxo-7-[2-(1H-1,2,3,4-tetrazol-1-yl)acetamido]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
Synonyms: sodium 3-{[(5-methyl-1,3,4-thiadiazol-2-yl)sulfanyl]methyl}-8-oxo-7-[2-(1,2,3,4-tetrazol-1-yl)acetamido]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
Molecular Weight (g/mol): 476.48
Molecular Formula: C14H13N8NaO4S3
InChi Key: FLKYBGKDCCEQQM-UHFFFAOYNA-M
SMILES: [Na+].CC1=NN=C(SCC2=C(N3C(SC2)C(NC(=O)CN2C=NN=N2)C3=O)C([O-])=O)S1
Physical Form 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Color 
  • (3)
Type 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)
  • (2)
  • (1)