Learn More
Invitrogen™ CDP-Star™ Substrate (0.25 mM Ready-To-Use)
Description
The CDP-Star™ Substrate is a chemiluminescent alkaline phosphatase substrate, ready-to-use for protein or nucleic acid blotting on nitrocellulose membranes. This substrate can also be used in solution-based assays.
This CDP-Star™ chemiluminescent substrate for alkaline phosphatase let you detect alkaline phosphatase and alkaline phosphatase-labeled molecules with unparalleled sensitivity, speed, and ease.
- Chemiluminescent substrate provides highly sensitive replacements for the widely used fluorogenic substrate methylumbelliferyl phosphate (MUP), and the colorimetric substrate p-nitrophenyl phosphate (pNPP).
- Low background luminescence coupled with high-intensity light output enables detection of alkaline phosphatase labels with the highest possible sensitivity and signal-to-noise.
This versatile chemiluminescent substrate exhibits high sensitivity in membrane-based applications such as Southern, Northern, and Western blotting. This substrate can also be used in solution-based assays such as immunoassays, DNA probe assays, enzyme assays, and reporter gene assays. Maximum light levels are reached in approximately 10 minutes and glow emission persists for several hours.
Specifications
Specifications
| Product Type | ELISA Substrate |
| Quantity | 100 mL |
| Detection Method | Chemiluminescence |
| Form | Liquid |
| Format | Bottle |
| Product Line | CDP-Star, NovaBright |
| Substrate Type | Alkaline Phosphatase |
| Content And Storage | Store the product at 4°C. |
| Shipping Condition | Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.