missing translation for 'onlineSavingsMsg'
Learn More

CDP-ethanolamine sodium salt hydrate, ≥93% (HPLC), MilliporeSigma™ Supelco™

$589.67 - $2346.86

Specifications

Linear Formula C11H20N4O11P2 · xNa+ · yH2 O
Synonym Cytidine 5′-diphosphate ethanolamine sodium salt hydrate; Cytidine 5′-diphosphoethanolamine sodium salt hydrate; Cytidine diphosphate ethanolamine sodium salt hydrate
Type Organic Standard
Formula Weight 446.24 (anhydrous free acid basis)
Percent Purity ≥93% (HPLC)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
111003851
View Documents
Merck Emd Millipore
9075610MG
10 mg
Each for $589.67
Only null left
 
111003049
View Documents
Merck Emd Millipore
9075650MG
50 mg
Each for $2,346.86
Only null left
 
Description

Description

Specifications

Specifications

C11H20N4O11P2 · xNa+ · yH2 O
Organic Standard
≥93% (HPLC)
Cytidine 5′-diphosphate ethanolamine sodium salt hydrate; Cytidine 5′-diphosphoethanolamine sodium salt hydrate; Cytidine diphosphate ethanolamine sodium salt hydrate
446.24 (anhydrous free acid basis)
Videos
SDS
Documents

Documents