missing translation for 'onlineSavingsMsg'
Learn More

CDP-ethanolamine sodium salt hydrate, ≥93% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111003851
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111003851 10 mg
111003049 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111003851 Supplier Merck Emd Millipore Supplier No. 9075610MG
Only null left

Specifications

Linear Formula C11H20N4O11P2 · xNa+ · yH2 O
Synonym Cytidine 5′-diphosphate ethanolamine sodium salt hydrate; Cytidine 5′-diphosphoethanolamine sodium salt hydrate; Cytidine diphosphate ethanolamine sodium salt hydrate
Type Organic Standard
Quantity 10 mg
Formula Weight 446.24 (anhydrous free acid basis)
Percent Purity ≥93% (HPLC)