missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CDK7/9tide Substrate

Catalog No. PV5090
Encompass
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PV5090 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PV5090 Supplier Thermo Scientific™ Supplier No. PV5090

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A peptide substrate based upon the C-terminal tail of RNA Polymerase II that is a reported substrate for Cyclin Dependent Kinases (CDK7, CDK8 and CDK9).

A peptide substrate based upon the C-terminal tail of RNA Polymerase II that is a reported substrate for Cyclin Dependent Kinases (CDK7, CDK8 and CDK9). For use with the Adapta™ Universal Kinase Assay (Cat. No. PV5099).

Specifications

Amino Acid Sequence YSPTSPSYSPTSPSYSPTSPSKKKK
Assay Entry Biochemical ADP accumulation
Chromagen Quantity 1mL
Content And Storage 2 mM CDK7/9tide peptide substrate in 50 mM HEPES (pH 7.5), 0.01% Brij-35, 10 mM MgCl2 and 1 mM EGTA.

Store at -20°C.
Detection Method Fluorescence
For Use With (Application) Pharma and biopharma, protein assays and analysis, target and lead identification and validation, Adapta™ Universal Kinase Assay (PV5099), drug discovery and development, kinase biology, biochemical kinase assays, Adapta™ universal kinase assays, enzyme and protein activity assays, protein assays and analysis, kinase activity
Quantity 1 mL
Substrate peptide based upon the C-terminal tail of RNA Polymerase II
Substrate Properties Peptide-Based Substrate
Substrate Type Kinase Substrate
Target Enzyme Serine/Threonine Kinases
Gene ID (Entrez) 1022, 3376
Molecular Weight (g/mol) 2689.9 Da
Product Type CDK7/9tide Substrate
Shipping Condition Dry Ice
Storage Requirements –20°C
Target Entry CDK9/Cyclin T1, CDK7
Validated Kinases CDK7, CDK9
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.