Learn More
Invitrogen™ Human CDK2/cyclin E1, GST Tag Recombinant Protein
Recombinant Protein
Supplier: Invitrogen™ PV6296
Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Recombinant full-length human CDK2 and CyclinE1 were co-expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag on both proteins. , Validated in LanthaScreen™ Kinase Binding Assay, Protocols available for simple use and optimized performance, Multiple pack sizes available for convenient use Protein Form: Recombinant The gene accession number for the subunits CKD2 and Cyclin E1 are NP_001789.2 and NP_001229.1.
CDK2/cyclinO is a member of the cyclin dependent protein kinase family complexed to CyclinO. CyclinO was used as a specific protein isoform name for the UNG gene which is localized exclusively in the nucleus. In addition, CyclinO was reported to shows significant uracil-DNA glycosylase activity. CDK2/cyclinO protein levels increase during G1 phase and the protein is turned over during the course of cell cycle. The levels of cyclin A-cdk2 is maximal at the G1/S transition and both cdk2 and cyclin A are found associated with DNA in the initiation complex during replication. Rb protein acts as substrate for cdk2-cyclin E and/or cdk2-cyclin A in vivo. Cdk2 is activated and specifically localized to the nucleus during late G1-, S-, and G2-phase.Specifications
| P24941 | |
| Kinase Assay | |
| 1017 | |
| Human CDK2/cyclin E1, GST Tag | |
| A630093N05Rik; CDC28; CDC2A; cdc2-related protein kinase; CDK1; CDK2; CDKN2; Cell division protein kinase 2; cyclin dependent kinase 2; cyclin dependent kinase 2-alpha; cyclin-dependent kinase 2; EC 2.7.11.22; EC 2.7.11.23; kinase Cdc2; MPF; p33 protein kinase; p33(CDK2); p33CDK2; p34 protein kinase | |
| 1-298 | |
| Human | |
| GST-tag | |
| Baculovirus |
| See Label | |
| 50 mM tris HCl with 0.1 mM EDTA, 0.1 mM PMSF, 0.25 mM DTT, 10 mM glutathione, 150 mM NaCl, 25% glycerol and no preservative; pH 7.5 | |
| 60.2-73.4 kDa | |
| 100 μg | |
| CDK2 | |
| Unconjugated | |
| Recombinant | |
| -80°C, Avoid Freeze/Thaw Cycles | |
| Liquid |