missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD56 Monoclonal Antibody (MEM-188), PE, Invitrogen™
Mouse Monoclonal Antibody
$1,754.00
Specifications
| Antigen | CD56 |
|---|---|
| Clone | MEM-188 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | PE |
Description
R-phycoerythrin (PE) is a stable and highly soluble phycobiliprotein which provides maximal absorbance and fluorescence without susceptibility to internal or external fluorescence quenching, thus providing an exceptional quantum yields and molar extinction coefficients.
CD56 Monoclonal antibody specifically detects CD56 in Human samples. It is validated for Flow Cytometry.Specifications
| CD56 | |
| Flow Cytometry | |
| PE | |
| Mouse | |
| PBS with 4mg/mL BSA, 10% sucrose and 0.1% sodium azide | |
| adhesion molecule; antigen recognized by monoclonal antibody 5.1H11; Cd56; CD-56; CD56 120 kDa GPI-linked isoform; CD56 140 kDa isoform; CD56 140 kDa VASE isoform; E NCAM; E-NCAM; MSK39; N CAM1; NCAM; N-CAM; Ncam1; N-CAM-1; NCAM-1; NCAMC; NCAM-C; neural cell adhesion molecule; Neural cell adhesion molecule 1; neural cell adhesion molecule, NCAM; sCD56; sNCAM; soluble CD56; soluble NCAM | |
| Ncam1 | |
| IgG2a κ | |
| Purified | |
| Antibody |
| MEM-188 | |
| Monoclonal | |
| Liquid | |
| Human | |
| P13591 | |
| 4684 | |
| Human CD56. | |
| Primary | |
| 4°C, store in dark | |
| Ncam1 |