missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD56 Monoclonal Antibody (MEM-188), PE, Invitrogen™
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MHCD56044
Description
R-phycoerythrin (PE) is a stable and highly soluble phycobiliprotein which provides maximal absorbance and fluorescence without susceptibility to internal or external fluorescence quenching, thus providing an exceptional quantum yields and molar extinction coefficients.
CD56, also known as neural cell adhesion molecule (NCAM), is a highly glycosylated transmembrane glycoprotein of the immunoglobulin family. It plays a crucial role in cell adhesion, migration, axonal growth, pathfinding, and synaptic plasticity. CD56 is ubiquitously expressed in the nervous system in isoforms ranging from 120-180 kDa and is involved in homotypic adhesion of neural cells. It mediates interactions by binding extracellular matrix components such as laminin and integrins, with polysialic modification reducing CD56-mediated adhesion. In the hematopoietic system, CD56 is expressed on natural killer (NK) cells and a subset of T cells known as NKT cells. It is also found on most neuroectodermal-derived cell lines, tissues, and neoplasms, including retinoblastoma, medulloblastoma, astrocytomas, and neuroblastoma. CD56 serves as a widely used neuroendocrine marker with high sensitivity for neuroendocrine tumors and ovarian granulosa cell tumors. Diseases associated with CD56 dysfunction include rabies and blastic plasmacytoid dendritic cell neoplasms, highlighting its importance in both neural and immune system functions.Chemical Identifiers
| CD56 | |
| Flow Cytometry | |
| PE | |
| Mouse | |
| PBS with 4mg/mL BSA, 10% sucrose and 0.1% sodium azide | |
| adhesion molecule; antigen recognized by monoclonal antibody 5.1H11; Cd56; CD-56; CD56 120 kDa GPI-linked isoform; CD56 140 kDa isoform; CD56 140 kDa VASE isoform; E NCAM; E-NCAM; MSK39; N CAM1; NCAM; N-CAM; Ncam1; N-CAM-1; NCAM-1; NCAMC; NCAM-C; neural cell adhesion molecule; Neural cell adhesion molecule 1; neural cell adhesion molecule, NCAM; sCD56; sNCAM; soluble CD56; soluble NCAM | |
| Ncam1 | |
| IgG2a κ | |
| Purified | |
| Antibody |
| MEM-188 | |
| Monoclonal | |
| Liquid | |
| Human | |
| P13591 | |
| 4684 | |
| Human CD56. | |
| Primary | |
| 4°C, store in dark | |
| Ncam1 |
Specifications
| CD56 | |
| Monoclonal | |
| PE | |
| Ncam1 | |
| adhesion molecule; antigen recognized by monoclonal antibody 5.1H11; Cd56; CD-56; CD56 120 kDa GPI-linked isoform; CD56 140 kDa isoform; CD56 140 kDa VASE isoform; E NCAM; E-NCAM; MSK39; N CAM1; NCAM; N-CAM; Ncam1; N-CAM-1; NCAM-1; NCAMC; NCAM-C; neural cell adhesion molecule; Neural cell adhesion molecule 1; neural cell adhesion molecule, NCAM; sCD56; sNCAM; soluble CD56; soluble NCAM | |
| Mouse | |
| Purified | |
| ASR, ASR/IVD | |
| 4684 | |
| 4°C, store in dark | |
| Liquid |
| Flow Cytometry | |
| MEM-188 | |
| PBS with 4mg/mL BSA, 10% sucrose and 0.1% sodium azide | |
| P13591 | |
| Ncam1 | |
| Human CD56. | |
| 2 mL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |