missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD249 Recombinant Rabbit Monoclonal Antibody (23GB5325)

Rabbit Recombinant Monoclonal Antibody
$656.94 - $1,779.35
Specifications
| Antigen | CD249 |
|---|---|
| Clone | 23GB5325 |
| Concentration | 1.5 mg/mL |
| Content And Storage | -20°C |
| Applications | Western Blot, Immunocytochemistry |
Description
CD249 Recombinant Monoclonal Antibody for ICC/IF, Western Blot
CD249 appears to have a role in the catabolic pathway of the renin-angiotensin system. Probably plays a role in regulating growth and differentiation of early B-lineage cells.Specifications
| CD249 | |
| 1.5 mg/mL | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P16406, P50123, Q07075 | |
| 13809, 2028, 64017 | |
| A synthesized peptide derived from human Aminopeptidase A/CD249 (900-957AA). | |
| Antibody |
| 23GB5325 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| Enpep | |
| 6030431M22Rik; Aminopeptidase A; aminopeptidase-A; APA; AP-A; BP1; Bp-1/6C3; BP-1/6C3 antigen; CD249; differentiation antigen gp160; EAP; ENPEP; Glutamyl aminopeptidase; glutamyl aminopeptidase (aminopeptidase A); gp160; Ly51; Ly-51 | |
| Enpep | |
| Primary | |
| Affinity chromatography |