missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD249 Recombinant Rabbit Monoclonal Antibody (23GB5325)

Catalog No. MA553887
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553887 20 μL
MA553886 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553887 Supplier Invitrogen™ Supplier No. MA553887
Only null left

Rabbit Recombinant Monoclonal Antibody

CD249 Recombinant Monoclonal Antibody for ICC/IF, Western Blot

CD249 appears to have a role in the catabolic pathway of the renin-angiotensin system. Probably plays a role in regulating growth and differentiation of early B-lineage cells.

Specifications

Antigen CD249
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB5325
Concentration 1.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Enpep
Gene Accession No. P16406, P50123, Q07075
Gene Alias 6030431M22Rik; Aminopeptidase A; aminopeptidase-A; APA; AP-A; BP1; Bp-1/6C3; BP-1/6C3 antigen; CD249; differentiation antigen gp160; EAP; ENPEP; Glutamyl aminopeptidase; glutamyl aminopeptidase (aminopeptidase A); gp160; Ly51; Ly-51
Gene Symbols Enpep
Host Species Rabbit
Immunogen A synthesized peptide derived from human Aminopeptidase A/CD249 (900-957AA).
Purification Method Affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13809, 2028, 64017
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less