missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ CD22L-N3 (BPC-Sia-LacNAc-N3)
Description
CD22L-N3 (BPC-Sia-LacNAc-N3) is a functionalized CD22 receptor ligand for trogocytosis targeting chimera (TrogoTAC) research and development. CD22 targeted TrogoTACs are bispecific molecules capable of inducing protein transfer from the target cell to B cells, by redirecting trogocytosis. Comprises an azide functionalized CD22 ligand for conjugation.
Specifications
Specifications
| Chemical Name or Material | (2R,4S,5R,6R)-6-((1R,2R)-3-([1,1'-Biphenyl]-4-carboxamido)-1,2-dihydroxypropyl)-5-acetamido-2-(((2R,3R,4S,5R,6S)-6-(((2R,3S,4R,5R,6R)-5-acetamido-6-(2-azidoethoxy)-4-hydroxy-2-(hydroxymethyl)tetrahydro-2H-pyran-3-yl)oxy)-3,4,5-trihydroxytetrahydro-2H |
| CAS | 1019642-47-7 |
| Quantity | 1 mg |
| Target | Glycobiology Probes |
| Molecular Formula | C40H54N6O19 |
| Purity | 95% |