Learn More
Gibco™ CD OptiCHO™ AGT™ Medium
CD OptiCHO™ AGT™ Medium
Supplier: Gibco™ A1122201
Description
CD OptiCHO™ medium is specifically designed to offer high performance and yield with recombinant CHO cells in a chemically defined fed-batch environment. The protein-free, animal origin free formulation is designed to provide consistency and reduces the need to screen for adventitious agents. CD OptiCHO™ medium builds on Gibco™ CD CHO's legacy of consistent cell growth and titers, while offering up to 40% better titer and cell growth over select competitors. Thanks to its low osmolality and advanced CHO CD EfficientFeed™ Liquid Nutrient Supplement Kit (Cat. No. A1024101) feeding options, CD OptiCHO™ medium is capable of gaining 2-5 fold increases in fed-batch productivity over batch culture.
When to Use CD OptiCHO™ Medium
- You are using a transfected CHO cell line other than CHO K1, GS CHO or CHO-S™ cell lines
- Optimization of feeding strategies will be part of your base medium selection testing in an effort to maximize productivity
- Your CHO cell line is 'finicky,' i.e. hard to grow or adapt
When to Consider Another Gibco™ Medium
Consider CD FortiCHO™ medium (Cat. No. A1148301) when:
- The cell line you are using is a transfected CHO K1, GS CHO or CHO-S™ cell line
- Maximum batch culture cell densities and protein titers are needed
- You have time to adapt cells into a new medium in an effort to maximize titers
- Cell health at harvest is a priority for downstream processing
Consider CD CHO medium (Cat. No. 10743029) when:
- Your GS CHO cells are recommended to be grown in CD CHO medium
- Your cells have been successfully cultured in CD CHO medium in the past and your process is nearing a regulatory filing (DMF available)
Consider CD DG44 medium (Cat. No. 12610010) when:
- You use parental DG44 or DXB11 CHO cells (dhfr-)
Liquid Media Formulations
Gibco™ AGT™ media are also available in a liquid format (Cat. No. 12681011) that offers the same performance and formulation as the AGT™ media format. Gibco™ liquid media are manufactured animal origin-free with the same complete, serum-free, protein-free, chemically-defined formulation and performance as their AGT™ media counterparts.
Design Your Own Media Specific to Your Needs
Whether it's a bench-level experiment or large scale biomanufacturing process, we offer specialized formats, packaging, and quality levels specific to your needs. With Gibco™ cGMP Media Custom Services you can put the manufacturing power and years of technical experience of our R&D and Bioproduction Application Specialists to work for you.
Specifications
| CHO | |
| Powder (AGT) | |
| Serum-free | |
| High Glucose, Sodium Pyruvate, Sodium Bicarbonate | |
| AGT, Gibco | |
| Wet Ice | |
| Ropak Bucket | |
| Magnesium, Sodium Phosphate | |
| 100 L | |
| 1 x 100 L, Storage 2-8°C Dark and Dry | |
| Suspension Cell Culture |
| Animal Origin-free, Chemically-defined, Protein-free, Serum-free | |
| CD OptiCHO AGT Medium | |
| Non-sterile | |
| No Glutamine, No Phenol Red | |
| 100 L | |
| Low | |
| No antibiotics | |
| Certified | |
| Medium does not support wild type DG44 cells | |
| CO2 |
For Research Use Only. Not for use in diagnostic procedures.