missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CCL1 Monoclonal Antibody (K56011_8A7), Invitrogen™
Mouse Monoclonal Antibody
$1,461.54 - $2,194.06
Specifications
| Antigen | CCL1 |
|---|---|
| Clone | K56011_8A7 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
Sequence of this protein is as follows: KSMQVPFSRC CFSFAEQEIP LRAILCYRNT SSICSNEGLI FKLKRGKEAC ALDTVGWVQR HRKMLRHCPS KRK
CCL1 Monoclonal antibody specifically detects CCL1 in Human samples. It is validated for Western BlotSpecifications
| CCL1 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| BF534335; C-C motif chemokine 1; C-C motif chemokine ligand 1; CCL 1; Ccl1; CCR8 ligand; chemokine (C-C motif) ligand 1; I-309; inflammatory cytokine I-309; P500; SCYA1; SISe; SIS-epsilon; small inducible cytokine A1; small inducible cytokine A1 (I-309, homologous to mouse Tca-3); small-inducible cytokine A1; T lymphocyte-secreted protein I-309; TCA3; TCA-3; T-cell activation protein 3 | |
| Recombinant human CCL1 (immunogen range is 24-96). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| CCL1 |
| K56011_8A7 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| P22362 | |
| 6346 | |
| IgG1 | |
| Protein A | |
| Antibody |