missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CCL1 Monoclonal Antibody (K56011_8A7), Invitrogen™

Catalog No. MA555739
Change view
Click to view available options
Quantity:
50 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA555739 50 μL
MA556119 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA555739 Supplier Invitrogen™ Supplier No. MA555739
Only null left

Mouse Monoclonal Antibody

Sequence of this protein is as follows: KSMQVPFSRC CFSFAEQEIP LRAILCYRNT SSICSNEGLI FKLKRGKEAC ALDTVGWVQR HRKMLRHCPS KRK

This gene is one of several cytokine genes clustered on the q-arm of chromosome 17. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The protein encoded by this gene is structurally related to the CXC subfamily of cytokines. Members of this subfamily are characterized by two cysteines separated by a single amino acid. This cytokine is secreted by activated T cells and displays chemotactic activity for monocytes but not for neutrophils. It binds to the chemokine receptor CCR8.

Specifications

Antigen CCL1
Applications Western Blot
Classification Monoclonal
Clone K56011_8A7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene CCL1
Gene Accession No. P22362
Gene Alias BF534335; C-C motif chemokine 1; C-C motif chemokine ligand 1; CCL 1; Ccl1; CCR8 ligand; chemokine (C-C motif) ligand 1; I-309; inflammatory cytokine I-309; P500; SCYA1; SISe; SIS-epsilon; small inducible cytokine A1; small inducible cytokine A1 (I-309, homologous to mouse Tca-3); small-inducible cytokine A1; T lymphocyte-secreted protein I-309; TCA3; TCA-3; T-cell activation protein 3
Gene Symbols CCL1
Host Species Mouse
Immunogen Recombinant human CCL1 (immunogen range is 24-96).
Purification Method Protein A
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6346
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less