missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCG-1423 98.0+%, TCI America™
$301.98 - $902.21
Chemical Identifiers
| CAS | 285986-88-1 |
|---|---|
| Molecular Formula | C18H13ClF6N2O3 |
| Molecular Weight (g/mol) | 454.753 |
| MDL Number | MFCD01566719 |
| InChI Key | DSMXVSGJIDFLKP-UHFFFAOYSA-N |
| Synonym | N-[2-[(4-Chlorophenyl)amino]-1-methyl-2-oxoethoxy]-3,5-bis(trifluoromethyl)benzamide |
| PubChem CID | 2726015 |
| IUPAC Name | N-[1-(4-chloroanilino)-1-oxopropan-2-yl]oxy-3,5-bis(trifluoromethyl)benzamide |
| SMILES | CC(C(=O)NC1=CC=C(C=C1)Cl)ONC(=O)C2=CC(=CC(=C2)C(F)(F)F)C(F)(F)F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
C344925MG
|
TCI America
C344925MG |
25 mg |
Each for $301.98
|
|
|||||
|
C3449100MG
|
TCI America
C3449100MG |
100 mg |
Each for $902.21
|
|
|||||
Chemical Identifiers
| 285986-88-1 | |
| 454.753 | |
| DSMXVSGJIDFLKP-UHFFFAOYSA-N | |
| 2726015 | |
| CC(C(=O)NC1=CC=C(C=C1)Cl)ONC(=O)C2=CC(=CC(=C2)C(F)(F)F)C(F)(F)F |
| C18H13ClF6N2O3 | |
| MFCD01566719 | |
| N-[2-[(4-Chlorophenyl)amino]-1-methyl-2-oxoethoxy]-3,5-bis(trifluoromethyl)benzamide | |
| N-[1-(4-chloroanilino)-1-oxopropan-2-yl]oxy-3,5-bis(trifluoromethyl)benzamide |
Specifications
| 285986-88-1 | |
| C18H13ClF6N2O3 | |
| 25 mg | |
| DSMXVSGJIDFLKP-UHFFFAOYSA-N | |
| N-[1-(4-chloroanilino)-1-oxopropan-2-yl]oxy-3,5-bis(trifluoromethyl)benzamide | |
| 2726015 | |
| ≥98.0% (GC) | |
| CCG-1423 |
| White-Yellow | |
| MFCD01566719 | |
| N-[2-[(4-Chlorophenyl)amino]-1-methyl-2-oxoethoxy]-3,5-bis(trifluoromethyl)benzamide | |
| CC(C(=O)NC1=CC=C(C=C1)Cl)ONC(=O)C2=CC(=CC(=C2)C(F)(F)F)C(F)(F)F | |
| 454.753 | |
| 454.75 | |
| Crystalline Powder |
Safety and Handling
TSCA : No
Recommended Storage : Freezer