missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| CCG-1423 | |
| White-Yellow | |
| MFCD01566719 | |
| N-[2-[(4-Chlorophenyl)amino]-1-methyl-2-oxoethoxy]-3,5-bis(trifluoromethyl)benzamide | |
| CC(C(=O)NC1=CC=C(C=C1)Cl)ONC(=O)C2=CC(=CC(=C2)C(F)(F)F)C(F)(F)F | |
| 454.753 | |
| 454.75 | |
| Crystalline Powder |
| 285986-88-1 | |
| C18H13ClF6N2O3 | |
| 100 mg | |
| DSMXVSGJIDFLKP-UHFFFAOYSA-N | |
| N-[1-(4-chloroanilino)-1-oxopropan-2-yl]oxy-3,5-bis(trifluoromethyl)benzamide | |
| 2726015 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 285986-88-1 | |
| 454.753 | |
| DSMXVSGJIDFLKP-UHFFFAOYSA-N | |
| 2726015 | |
| CC(C(=O)NC1=CC=C(C=C1)Cl)ONC(=O)C2=CC(=CC(=C2)C(F)(F)F)C(F)(F)F |
| C18H13ClF6N2O3 | |
| MFCD01566719 | |
| N-[2-[(4-Chlorophenyl)amino]-1-methyl-2-oxoethoxy]-3,5-bis(trifluoromethyl)benzamide | |
| N-[1-(4-chloroanilino)-1-oxopropan-2-yl]oxy-3,5-bis(trifluoromethyl)benzamide |
Safety and Handling
TSCA : No
Recommended Storage : Freezer