missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCF4-AM
Description
CCF4-AM, our LiveBLAzer™ substrate, is a lipophilic, esterified form of the CCF4 substrate which allows it to readily enter cells. Upon entry, cleavage by endogenous cytoplasmic esterases rapidly converts CCF4-AM into its negatively charged form, CCF4, which is retained in the cytosol. CCF4 is a Fluorescence Resonance Energy Transfer (FRET) substrate which consists of a cephalosporin core linking 7-hydroxycoumarin to fluorescein. This product is to be used as a FRET-based substrate for β-lactamase as a sensitive reporter of mammalian gene expression in the GeneBLAzer™ technology.
Specifications
Specifications
| Cell Permeability | Cell-Permeant |
| Content And Storage | Store in freezer (-5 to -30°C). |
| Detection Method | Fluorescence |
| Form | Solid |
| Product Line | LiveBLAzer |
| Quantity | 20mg |
| Substrate | CCF4-AM |
| Substrate Properties | Chemical Substrate |
| Substrate Type | Beta-Lactamase Substrate |
| For Use With (Equipment) | Flow Cytometer, Microplate Reader |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.