missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CCF4-AM

Catalog No. K1028
Encompass
Change view
Click to view available options
Quantity:
5mg
20mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K1028 5mg
K1029 20mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. K1028 Supplier Invitrogen™ Supplier No. K1028
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CCF4-AM

CCF4-AM, our LiveBLAzer™ substrate, is a lipophilic, esterified form of the CCF4 substrate which allows it to readily enter cells. Upon entry, cleavage by endogenous cytoplasmic esterases rapidly converts CCF4-AM into its negatively charged form, CCF4, which is retained in the cytosol. CCF4 is a Fluorescence Resonance Energy Transfer (FRET) substrate which consists of a cephalosporin core linking 7-hydroxycoumarin to fluorescein. This product is to be used as a FRET-based substrate for β-lactamase as a sensitive reporter of mammalian gene expression in the GeneBLAzer™ technology.

Specifications

Cell Permeability Cell-Permeant
Content And Storage Store in freezer (-5 to -30°C).
Detection Method Fluorescence
Form Solid
Product Line GeneBLAzer
Quantity 5mg
Substrate CCF4-AM
Substrate Properties Chemical Substrate
Substrate Type Beta-Lactamase Substrate
For Use With (Equipment) Flow Cytometer, Microplate Reader
Shipping Condition Room Temperature
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.