missing translation for 'onlineSavingsMsg'
Learn More

CCF2-FA (free acid)

Catalog No. K1027
Encompass
Change view
Click to view available options
Quantity:
100μg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K1027 1mg
K1034 100μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. K1027 Supplier Thermo Fisher Scientific Supplier No. K1027

CCF2-FA (free acid)

CCF2-FA (Free Acid) is a fluorescence resonance energy transfer (FRET) substrate that consists of a cephalosporin core linking B7-hydroxycoumarin to fluorescein. This product is intended for use as a beta-lactamase substrate in a GeneBLAzer™ cell-based assay.

GeneBLAzer™ cell-based assays typically utilize the membrane-permeant ester forms (CCF2-AM and CCF4-AM) of the negatively charged fluorescent beta-lactamase substrates, CCF2 and CCF4. These lipophilic esters readily enter the cell, where cleavage by endogeneous cytoplasmic esterases rapidly converts them into their negatively charged forms, thereby trapping them in the cytosol.

CCF2-FA is essentially the CCF2 substrate without the esters found in the AM version. CCF2-FA is de-esterified and used in cell lysate applications, bypassing loading across the cell membrane and de-esterification steps. Cell lysates are the preferred method for applications using cells that contain a cell wall. CCF2-FA can also be used as a control to acquire the excitation and emission spectra for CCF2-AM and CCF4-AM.

Specifications

Cell Permeability Cell-Impermeant
Content And Storage Store in freezer (-5 to -30°C).
Detection Method Fluorescence
Product Line GeneBLAzer
Quantity 1mg
Substrate CCF2-FA
Substrate Properties Chemical Substrate
Substrate Type Beta-Lactamase Substrate
Shipping Condition Approved for shipment on Wet or Dry Ice

For Research Use Only. Not for use in diagnostic procedures.