missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ CBA
Description
CBA is a selective TRPM4 blocker (IC50 = 1.5 μM). Exhibits no significant activity against TRPM5, TRPM7, TRPM8, TRPV1, TRPV3, TRPV6 and a range of other ion channels and receptors. Displays neuroprotective effects against glutamate-induced neurodegeneration in vitro.
Specifications
Specifications
| Chemical Name or Material | 4-Chloro-2-[[2-(2-chlorophenoxy)acetyl]amino]benzoic acid |
| CAS | 351424-20-9 |
| Quantity | 10 mg |
| Molecular Formula | C15H11Cl2NO4 |
| Formula Weight | 340.16 |
| Purity | 98% |