missing translation for 'onlineSavingsMsg'
Learn More

(-)-Caryophyllene oxide, 95%

CAS: 1139-30-6 | C15H24O | 220.35 g/mol

Supplier:  Thermo Scientific Chemicals 325080500

 View more versions of this product

Catalog No. AC325080500


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

(-)-Caryophyllene oxide
94.0
55°C to 62°C
0.9600g/mL
Authentic
Glass bottle
1.4956
50 g
0.96
caryophyllene oxide, --caryophyllene oxide, beta-caryophyllene epoxide, beta-caryophyllene oxide, --epoxycaryophyllene, unii-s2xu9k448u, -carophyllene oxide, caryophyllene epoxide, epoxycaryophyllene, trans-caryophyllene oxide
CC1(CC2C1CCC3(C(O3)CCC2=C)C)C
1742210
220.35
Crystalline Powder
1139-30-6
100.0
White
>110°C
94% min. (GC)
C15H24O
MFCD00134216
17, IV, 392
-55
NVEQFIOZRFFVFW-RGCMKSIDSA-N
220.35
CHEBI:67818
95%

Chemical Identifiers

1139-30-6
220.35
NVEQFIOZRFFVFW-RGCMKSIDSA-N
1742210
CC1(CC2C1CCC3(C(O3)CCC2=C)C)C
C15H24O
MFCD00134216
caryophyllene oxide, --caryophyllene oxide, beta-caryophyllene epoxide, beta-caryophyllene oxide, --epoxycaryophyllene, unii-s2xu9k448u, -carophyllene oxide, caryophyllene epoxide, epoxycaryophyllene, trans-caryophyllene oxide
CHEBI:67818
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic to aquatic life with long lasting effects.

GHS P Statement
Avoid release to the environment.
Collect spillage.
Dispose of contents/container to an approved waste disposal plant.

EINECSNumber : 214-519-7

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only