accessibility menu, dialog, popup

UserName

CAS RN 1139-30-6

IUPAC Name: 4,12,12-trimethyl-9-methylidene-5-oxatricyclo[8.2.0.0⁴,⁶]dodecane
Synonyms: caryophyllene oxide (-)-β-caryophyllene epoxide
Molecular Weight (g/mol): 220.36
Molecular Formula: C15H24O
InChi Key: NVEQFIOZRFFVFW-UHFFFAOYNA-N
SMILES: CC12CCC3C(CC3(C)C)C(=C)CCC1O2
Percent Purity 
  • (1)
  • (1)
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)