missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carrez II Solution, Fisher Chemical™
$161.00 - $170.97
Specifications
| Chemical Name or Material | Carrez II Solution |
|---|---|
| CAS | 7446-20-0 |
| CAS Min % | 91.0,6.59 |
| CAS Max % | 95.0,6.85 |
| Grade | Laboratory |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
SC9102-250
|
Ricca Chemical Company
SC9102250 |
250 mL |
Each for $161.00
|
|
|||||
|
SC9102500
|
Ricca Chemical Company
SC9102500 |
500 mL |
Each for $170.97
|
|
|||||
Specifications
| Carrez II Solution | |
| 91.0,6.59 | |
| Laboratory | |
| For AOAC Method 984.15 |
| 7446-20-0 | |
| 95.0,6.85 | |
| Natural Poly Bottle |