missing translation for 'onlineSavingsMsg'
Learn More

Carnosic Acid 97.0+%, TCI America™

Catalog No. p-7125972
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 mg
20 mg
Catalog No. Quantity
C347720MG 20 mg
C3477100MG 100 mg
2 options

Catalog No. C347720MG

Supplier: TCI America C347720MG

Only null left

Chemical Identifiers

CAS 3650-09-7
Molecular Formula C20H28O4
Molecular Weight (g/mol) 332.44
MDL Number MFCD02259459
InChI Key QRYRORQUOLYVBU-VBKZILBWSA-N
Synonym (4aR,10aS)-5,6-Dihydroxy-7-isopropyl-1,1-dimethyl-1,2,3,4,4a,9,10,10a-octahydrophenanthrene-4a-carboxylic Acid
PubChem CID 65126
ChEBI CHEBI:65585
IUPAC Name (4aR,10aS)-5,6-dihydroxy-1,1-dimethyl-7-propan-2-yl-2,3,4,9,10,10a-hexahydrophenanthrene-4a-carboxylic acid
SMILES CC(C)C1=C(C(=C2C(=C1)CCC3C2(CCCC3(C)C)C(=O)O)O)O

Specifications

Melting Point 190°C
Color Yellow
Quantity 20 mg
Formula Weight 332.44
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Carnosic Acid
TSCA No
Recommended Storage Freezer