accessibility menu, dialog, popup

UserName

CAS RN 3650-09-7

IUPAC Name: (4aR,10aS)-5,6-dihydroxy-1,1-dimethyl-7-(propan-2-yl)-1,2,3,4,4a,9,10,10a-octahydrophenanthrene-4a-carboxylic acid
Synonyms: salvin carnosic acid
Molecular Weight (g/mol): 332.44
Molecular Formula: C20H28O4
InChi Key: QRYRORQUOLYVBU-VBKZILBWSA-N
SMILES: CC(C)C1=CC2=C(C(O)=C1O)[C@]1(CCCC(C)(C)[C@@H]1CC2)C(O)=O
Quantity 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (4)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (2)
  • (3)
  • (2)
  • (2)
  • (4)
  • (1)