missing translation for 'onlineSavingsMsg'
Learn More

Carnosic Acid 97.0+%, TCI America™

Catalog No. C347720MG
Change view
Click to view available options
Quantity:
20 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C347720MG 20 mg
C3477100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C347720MG Supplier TCI America Supplier No. C347720MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3650-09-7
Molecular Formula C20H28O4
Molecular Weight (g/mol) 332.44
MDL Number MFCD02259459
InChI Key QRYRORQUOLYVBU-VBKZILBWSA-N
Synonym (4aR,10aS)-5,6-Dihydroxy-7-isopropyl-1,1-dimethyl-1,2,3,4,4a,9,10,10a-octahydrophenanthrene-4a-carboxylic Acid
PubChem CID 65126
ChEBI CHEBI:65585
IUPAC Name (4aR,10aS)-5,6-dihydroxy-1,1-dimethyl-7-propan-2-yl-2,3,4,9,10,10a-hexahydrophenanthrene-4a-carboxylic acid
SMILES CC(C)C1=C(C(=C2C(=C1)CCC3C2(CCCC3(C)C)C(=O)O)O)O

Specifications

Melting Point 190°C
Color Yellow
Quantity 20 mg
Formula Weight 332.44
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Carnosic Acid
TSCA No
Recommended Storage Freezer