missing translation for 'onlineSavingsMsg'
Learn More

Carfilzomib, Thermo Scientific™

Catalog No. AC469111000
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC469111000 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC469111000 Supplier Thermo Scientific Chemicals Supplier No. 469111000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 868540-17-4
Molecular Formula C40 H57 N5 O7

Specifications

Quantity 100 mg
Melting Point 207°C to 210°C
Formula Weight 712.92
Physical Form Crystalline Powder
Color White to Almost-white
Purity ≥97.5%
Chemical Name or Material Carfilzomib
Hazard Category Exclamation mark
Recommended Storage Normal conditions