missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ CarePac™ Small Weights with Certificate

Higher process safety for more efficient testing

Supplier:  Mettler Toledo™ 11123106

 View more versions of this product

Catalog No. 01-911-858


Only null left
Explore available promotions

Includes:

Calibration certificate, tweezers, gloves and other accessories for professional weight handling


Description

Description

Perform routine testing of balances securely with only what you need – two test weights.

  • Reduced measurement deviations
  • Process tolerances up to 0.03%; supported by manufacturer SOPs

Compliance

ISO17025

Specifications

Specifications

Small Weight Set
ASTM Class 1
1, 20 g
1 g & 20 g
2/Set
ISO17025
Stainless Steel
7.9kg/dm3 at 20°C
Two test weights: 1 and 20g, tweezers, gloves
Multiple
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions