missing translation for 'onlineSavingsMsg'
Learn More

Heathrow Scientific™ Cardboard Cryogenic Freezer Box Partition/Divider
GREENER_CHOICE

Box for storage of small samples and tubes.

$82.71 - $87.11

Specifications

Color White
Material Cardboard
Shape Square
Type Box Divider
For Use With (Equipment) 1.5/2.0 mL Cryogenic Vials
View More Specs

Products 4
Catalog Number Mfr. No. Holds Price Quantity & Availability  
Catalog Number Mfr. No. Holds Price Quantity & Availability  
17100450
GREENER_CHOICE
View Documents Promotion Details
Heathrow Scientific™
HS2860E
100 Cryogenic Vials
Pack of 12 Each for $87.11
Only null left
 
17100428
GREENER_CHOICE
View Documents Promotion Details
Heathrow Scientific™
HS2860F
64 Cryogenic Vials
Pack of 12 Each for $82.71
Only null left
 
17100459
GREENER_CHOICE
View Documents Promotion Details
Heathrow Scientific™
HS2860G
49 Cryogenic Vials
Pack of 12 Each for $82.71
Only null left
 
17100457
GREENER_CHOICE
View Documents Promotion Details
Heathrow Scientific™
HS2860H
81 Cryogenic Vials
Pack of 12 Each for $87.11
Only null left
 
Description

Description

  • Array: 10 x 10
  • Low Temp: -196°C
Specifications

Specifications

White
Square
1.5/2.0 mL Cryogenic Vials
Cardboard
Box Divider
-196°C
Videos
SDS
Documents

Documents