missing translation for 'onlineSavingsMsg'
Learn More

Carboxymethylcellulose sodium salt

CAS: 9004-32-4 | (C12 H14 O9 R6)n | 263.20 g/mol

Supplier:  Thermo Scientific Chemicals A181050E

Quantity 2500 g

 View more versions of this product

Catalog No. AAA181050E


Description

Description

Carboxymethylcellulose sodium salt is used in drilling muds, in detergents as a soil-suspending agent, in resin emulsion paints, adhesives, printing inks, textile sizes and protective colloid. It acts as a stabilizer in foods. It is also employed in pharmaceuticals as a suspending agent and excipients for tablets. It is used as viscosity modifiers to stabilize the emulsions. It is used as a lubricant in artificial tears and it is used to characterize enzyme activity from endoglucanases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Carboxymethylcellulose sodium salt is used in drilling muds, in detergents as a soil-suspending agent, in resin emulsion paints, adhesives, printing inks, textile sizes and protective colloid. It acts as a stabilizer in foods. It is also employed in pharmaceuticals as a suspending agent and excipients for tablets. It is used as viscosity modifiers to stabilize the emulsions. It is used as a lubricant in artificial tears and it is used to characterize enzyme activity from endoglucanases.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

9004-32-4
263.20
DPXJVFZANSGRMM-UHFFFAOYNA-N
23706213
[Na].CC(O)=O.OCC(O)C(O)C(O)C(O)C=O
(C12 H14 O9 R6)n
MFCD00081472
carboxymethylcellulose sodium usp, celluvisc tn, carmellose sodium jp17, sodium dextrose acetate, c.m.c. tn
sodium;2,3,4,5,6-pentahydroxyhexanal;acetate

Specifications

Carboxymethylcellulose sodium salt
>300°C
2500 g
14,1829
Soluble in water.
[Na].CC(O)=O.OCC(O)C(O)C(O)C(O)C=O
263.20
9004-32-4
(C12 H14 O9 R6)n
MFCD00081472
carboxymethylcellulose sodium usp, celluvisc tn, carmellose sodium jp17, sodium dextrose acetate, c.m.c. tn
DPXJVFZANSGRMM-UHFFFAOYNA-N
sodium;2,3,4,5,6-pentahydroxyhexanal;acetate
23706213
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions