missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carboxymethylcellulose Sodium, Low Viscosity, USP, Spectrum™ Chemical
CA193, 9004-32-4
Supplier: Spectrum Chemical Mfg Cor CA19325KG
Description
Spectrum™ Chemical Carboxymethylcellulose Sodium, Low Viscosity, Viscosity of 2 Percent Aqueous Solution @ 25 DEG C: 37.5 cP, USP is often used as a thickener and stabilizer. The USP grade indicates it is a grade suitable for personal care, cosmetic and pharmaceutical applications and were manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
| 9004-32-4 | |
| (C12 H14 O9 R6)n | |
| 2.5 kg | |
| [Na].CC(O)=O.OCC(O)C(O)C(O)C(O)C=O | |
| 263.20 | |
| 10% |
| 100% | |
| MFCD00081472 | |
| DPXJVFZANSGRMM-UHFFFAOYNA-N | |
| 2,3,4,5,6-pentahydroxyhexanal acetic acid sodium | |
| USP | |
| Poly Pail |
Chemical Identifiers
| 9004-32-4 | |
| 263.20 | |
| DPXJVFZANSGRMM-UHFFFAOYNA-N | |
| [Na].CC(O)=O.OCC(O)C(O)C(O)C(O)C=O |
| (C12 H14 O9 R6)n | |
| MFCD00081472 | |
| 2,3,4,5,6-pentahydroxyhexanal acetic acid sodium |