missing translation for 'onlineSavingsMsg'
Learn More

Carboxymethylcellulose Sodium, Low Viscosity, USP, Spectrum™ Chemical

Catalog No. p-7157249
Change view
Click to view available options
Quantity:
100 g
125 g
500 g
2.5 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18601480 100 g Poly Bottle
18601476 125 g Poly Bottle
18601477 500 g Poly Bottle
18601478 2.5 kg Poly Pail
18601479 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18601480 Supplier Spectrum Chemical Mfg Cor Supplier No. CA193100GM
Only null left

CA193, 9004-32-4

Spectrum™ Chemical Carboxymethylcellulose Sodium, Low Viscosity, Viscosity of 2 Percent Aqueous Solution @ 25 DEG C: 37.5 cP, USP is often used as a thickener and stabilizer. The USP grade indicates it is a grade suitable for personal care, cosmetic and pharmaceutical applications and were manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 9004-32-4
Molecular Formula (C12 H14 O9 R6)n
Molecular Weight (g/mol) 263.20
MDL Number MFCD00081472
InChI Key DPXJVFZANSGRMM-UHFFFAOYNA-N
IUPAC Name 2,3,4,5,6-pentahydroxyhexanal acetic acid sodium
SMILES [Na].CC(O)=O.OCC(O)C(O)C(O)C(O)C=O

Specifications

CAS Max % 100%
Quantity 100 g
Grade USP
Loss on Drying 10%
Packaging Poly Bottle