missing translation for 'onlineSavingsMsg'
Learn More

Carboxymethyl Cellulose Sodium Salt, Low Vicosity, MP Biomedicals

Catalog No. ICN10127891
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10127891 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10127891 Supplier MP Biomedicals Inc Supplier No. 0210127891

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 9004-32-4
Molecular Formula (C12 H14 O9 R6)n
Molecular Weight (g/mol) 263.20
MDL Number MFCD00081472
InChI Key DPXJVFZANSGRMM-UHFFFAOYNA-N
Synonym carboxymethylcellulose sodium usp, celluvisc tn, carmellose sodium jp17, sodium dextrose acetate, c.m.c. tn
PubChem CID 23706213
SMILES [Na].CC(O)=O.OCC(O)C(O)C(O)C(O)C=O

Specifications

Color White
Quantity 1 kg
IUPAC Name 2,3,4,5,6-pentahydroxyhexanal acetic acid sodium
Formula Weight ≈90 kd
Grade Not Available
Physical Form Powder
Recommended Storage Store at Room Temperature (15 to 30°C).