Learn More
MilliporeSigma™ Supelco™ Carboxen™ 572 SPE for Tobacco, Bed Wt. 300 mg
Solid phase extraction (SPE) is a technique commonly used in laboratories to isolate analytes of interest from complex matrices.
$1313.10 - $1313.10
Specifications
| Basis Weight | 300 mg |
|---|---|
| Material | Carboxen 572 |
| For Use With (Application) | Environmental and Industrial Hygiene |
| Product Line | Carboxen |
| Type | SPE Column |
Description
Rezorian Cartridges are made of polypropylene and typically contain one bed of adsorbent with polyethylene frits. The end fittings are luer lock syringe connections that can be used to connect to sampling pump tubing,Specifications
| 300 mg | |
| Environmental and Industrial Hygiene | |
| SPE Column |
| Carboxen 572 | |
| Carboxen | |
| 3 mL |
Carboxen is a registered trademark of Sigma-Aldrich Co. LLC