missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Carbosieve™ SIII Thermal Desorption Tubes

Catalog No. 111006749
Change view
Click to view available options
Description:
Pre-conditioned
Pre-conditioned/Sealed with Brass Endcaps
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
111006749 Pre-conditioned
111006687 Pre-conditioned/Sealed with Brass Endcaps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111006749 Supplier MilliporeSigma™ Supelco™ Supplier No. 20012U

Suitable for use with Air Monitoring

Specifications

Diameter (English) Outer 1/4 in.
Packaging Quantity 10/Pk.
Product Line Carbosieve
Overall Length (English) 3.5 in.
For Use With (Application) Air Monitoring
Matrix Carbosieve™ S-III Carbon Molecular Sieve
For Use With (Equipment) PerkinElmer, Markes, DANI, OI Analytical, and Shimadzu Instruments
Methods EPA TO-2
Type Desorption Tube
Description Pre-conditioned
Material Stainless Steel
Show More Show Less

Carbosieve is a registered trademark of Sigma-Aldrich Co. LLC