missing translation for 'onlineSavingsMsg'
Learn More

Carbonylhydridotris(triphenylphosphine)rhodium(I), TCI America™

Catalog No. C13831G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C13831G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C13831G Supplier TCI America Supplier No. C13831G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 17185-29-4
Molecular Formula C55H46OP3Rh
Molecular Weight (g/mol) 918.80
MDL Number MFCD00151644
InChI Key AACIZACVKFEETJ-UHFFFAOYSA-N
Synonym tristriphenylphosphine rhodium carbonyl hydride, hydridocarbonyltris triphenylphosphine rhodium i
PubChem CID 132451836
IUPAC Name methanidylidyneoxidanium tris(triphenylphosphane) λ¹-rhodium
SMILES [RhH].[C-]#[O+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 122°C
Color Yellow
Quantity 1 g
UN Number 2811
Formula Weight 918.80
Physical Form Crystalline Powder
Chemical Name or Material Carbonylhydridotris(triphenylphosphine)rhodium(I)