missing translation for 'onlineSavingsMsg'
Learn More

Carbonylchlorohydridotris(triphenylphosphine)ruthenium(II)

Catalog No. AC392140010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC392140010 1 g Glass bottle
AC392140050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC392140010 Supplier Thermo Scientific Chemicals Supplier No. 392140010

CAS: 16971-33-8 | C55H46ClOP3Ru | 952.41 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent

Chemical Identifiers

CAS 16971-33-8
Molecular Formula C55H46ClOP3Ru
Molecular Weight (g/mol) 952.41
MDL Number MFCD00049804
InChI Key IMPHYUNMDWWJAH-UHFFFAOYSA-M
Synonym carbonylchlorohydridotris triphenylphosphine-ruthenium ii
PubChem CID 71463878
IUPAC Name formaldehyde;hydride;ruthenium(2+);triphenylphosphane;chloride
SMILES Cl[RuH++].[C-]#[O+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 204.0°C to 206.0°C
Color Cream to Tan
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble
Formula Weight 952.4
Physical Form Powder
Chemical Name or Material Carbonylchlorohydridotris(triphenylphosphine)ruthenium(II)
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 241-051-

RUO – Research Use Only