missing translation for 'onlineSavingsMsg'
Learn More

Carbonyl Cyanide 4-(Trifluoromethoxy)phenylhydrazone 95.0+%, TCI America™

Catalog No. C346310MG
Change view
Click to view available options
Quantity:
10 mg
50 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C346310MG 10 mg
C346350MG 50 mg
C3463250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C346310MG Supplier TCI America Supplier No. C346310MG

Chemical Identifiers

CAS 370-86-5
Molecular Formula C10H5F3N4O
Molecular Weight (g/mol) 254.172
MDL Number MFCD00009699
InChI Key BMZRVOVNUMQTIN-UHFFFAOYSA-N
Synonym fccp, carbonyl cyanide p-trifluoromethoxyphenylhydrazone, carbonyl cyanide 4-trifluoromethoxy phenylhydrazone, 2-2-4-trifluoromethoxy phenyl hydrazono malononitrile, propanedinitrile, 4-trifluoromethoxy phenyl hydrazono, 1-cyano-n-4-trifluoromethoxy phenyl methanecarbohydrazonoyl cyanide, carbonyl cyanide 4-trifluoromethoxyphenylhydrazone, carbonyl cyanide, 4-trifluoromethoxyphenylhydrazone, 4-trifluoromethoxy phenyl hydrazono malononitrile, mesoxalonitrile, p-trifluoromethoxy phenyl hydrazone
PubChem CID 3330
ChEBI CHEBI:75458
IUPAC Name 2-[[4-(trifluoromethoxy)phenyl]hydrazinylidene]propanedinitrile
SMILES C1=CC(=CC=C1NN=C(C#N)C#N)OC(F)(F)F

Specifications

Melting Point 156°C
Color Yellow
Quantity 10 mg
UN Number 3439
Formula Weight 254.17
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Carbonyl Cyanide 4-(Trifluoromethoxy)phenylhydrazone